Learn More
Medchemexpress LLC HY-P7274 10ug Medchemexpress, OSM (HEK293-expressed), Mouse CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP727410UG
Encompass_Preferred
Medchemexpress, HY-P7274 10ug OSM (HEK293-expressed), Mouse CAS: Recombinant Mouse Oncostatin M (HEK293-expressed) is a gp130 cytokine family member, shows homeostatic and proinflammatory activities and can modify extracellular matrix. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3139