Learn More
Medchemexpress LLC OSM Human 209a.a 100ug
SDP
Catalog No. 5000184676
Encompass_Preferred
Recombinant human Oncostatin M (OSM) is a 209-amino-acid cytokine of the IL-6 family involved in hematopoiesis, mesenchymal stem cell differentiation, tissue regeneration, inflammation, and metabolism. This lyophilized recombinant protein is provided for research applications with recommended storage and handling instructions to preserve activity.
  • Contains full-length human OSM (209 amino acids).
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight 24-26 kDa under reducing conditions.
  • Lyophilized from PBS (pH 7.4) with 8% trehalose for stability.
  • Expressed in E. coli as a recombinant protein.
  • Stable when stored at -20 °C; aliquot and store at -80 °C for extended storage.
Catalog No. 50-001-84676 Supplier Medchemexpress LLC Supplier No. HYP7052A100UG
Recombinant human Oncostatin M (OSM) is a 209-amino-acid cytokine of the IL-6 family involved in hematopoiesis, mesenchymal stem cell differentiation, tissue regeneration, inflammation, and metabolism. This lyophilized recombinant protein is provided for research applications with recommended storage and handling instructions to preserve activity.
  • Contains full-length human OSM (209 amino acids).
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight 24-26 kDa under reducing conditions.
  • Lyophilized from PBS (pH 7.4) with 8% trehalose for stability.
  • Expressed in E. coli as a recombinant protein.
  • Stable when stored at -20 °C; aliquot and store at -80 °C for extended storage.