Learn More
Medchemexpress LLC OSM Human 209a.a 50ug

Supplier: Medchemexpress LLC HYP7052A50UG
Recombinant, tag-free human Oncostatin M (209 amino acids) supplied for research use. Oncostatin M is an IL-6 family cytokine involved in hematopoiesis, mesenchymal stem cell differentiation, liver regeneration, heart remodeling, nociception, inflammation, and metabolism.
- Recombinant, tag-free human Oncostatin M with 209 amino acids.
- Expressed in Escherichia coli to provide consistent product quality.
- Lyophilized formulation from 0.22 μm filtered PBS (pH 7.4) with 8% trehalose.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Stable at -20 °C for up to 2 years; reconstituted material is stable at 4 °C for 1 week or -20 °C for longer with carrier protein.
- Available in multiple sizes including small research amounts and larger quantities.
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.