Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC OSM Human 227a.a 10ug
SDP

Catalog No. 5000185047
Encompass_Preferred

Recombinant human Oncostatin M (OSM) is a full-length 227 amino acid cytokine of the IL-6 family supplied lyophilized for research applications. OSM modulates hematopoiesis, mesenchymal stem cell differentiation, liver regeneration, cardiac remodeling, nociception, inflammation, and metabolic pathways. The protein is expressed in E. coli, tag-free, and purified for reliable functional studies.

  • Full-length human sequence (227 amino acids).
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Expressed in E. coli to provide a non-glycosylated recombinant protein.
  • Tag-free preparation to minimize assay interference.
  • Lyophilized formulation with stabilizer for improved shelf life.
  • Suitable for cell signaling and functional assays involving OSM activity.
  • Storage recommendations provided for short- and long-term stability.

Catalog No. 50-001-85047 Supplier Medchemexpress LLC Supplier No. HYP7052B10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Oncostatin M (OSM) is a full-length 227 amino acid cytokine of the IL-6 family supplied lyophilized for research applications. OSM modulates hematopoiesis, mesenchymal stem cell differentiation, liver regeneration, cardiac remodeling, nociception, inflammation, and metabolic pathways. The protein is expressed in E. coli, tag-free, and purified for reliable functional studies.

  • Full-length human sequence (227 amino acids).
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Expressed in E. coli to provide a non-glycosylated recombinant protein.
  • Tag-free preparation to minimize assay interference.
  • Lyophilized formulation with stabilizer for improved shelf life.
  • Suitable for cell signaling and functional assays involving OSM activity.
  • Storage recommendations provided for short- and long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.