Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC OSM Human 227a.a 2ug
SDP

Catalog No. 5000185441
Encompass_Preferred

Recombinant human Oncostatin M (OSM) is a 227-amino-acid cytokine of the interleukin-6 family supplied as a lyophilized, tag-free recombinant protein produced in E. coli. It is provided at high purity for in vitro research applications including cell signaling, hematopoiesis, inflammation, and tissue remodeling, with recommended storage and handling guidance.

  • Recombinant, tag-free protein expressed in E. coli.
  • 227 amino acid sequence corresponding to human Oncostatin M.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation with stabilizer for shelf stability.
  • Includes accession and gene identifiers for reference.
  • Intended for research use with recommended storage conditions.

Catalog No. 50-001-85441 Supplier Medchemexpress LLC Supplier No. HYP70522UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Oncostatin M (OSM) is a 227-amino-acid cytokine of the interleukin-6 family supplied as a lyophilized, tag-free recombinant protein produced in E. coli. It is provided at high purity for in vitro research applications including cell signaling, hematopoiesis, inflammation, and tissue remodeling, with recommended storage and handling guidance.

  • Recombinant, tag-free protein expressed in E. coli.
  • 227 amino acid sequence corresponding to human Oncostatin M.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation with stabilizer for shelf stability.
  • Includes accession and gene identifiers for reference.
  • Intended for research use with recommended storage conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.