Learn More
Medchemexpress LLC OSM Human HEK293 100ug
SDP
Catalog No. 5000179822
Encompass_Preferred
OSM protein is a multifunctional growth regulator with dual functions of inhibiting tumor cell lines and stimulating the proliferation of AIDS-KS cells It coordinates the production of cytokines including IL-6 G-CSF and GM-CSF and binds to type I and type II OSM receptors forming heterodimers with LIFR/IL6ST and OSMR/IL6ST respectively OSM Protein Human (HEK293 His) is the recombinant human-derived OSM protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79822 Supplier Medchemexpress LLC Supplier No. HYP73331100UG
OSM protein is a multifunctional growth regulator with dual functions of inhibiting tumor cell lines and stimulating the proliferation of AIDS-KS cells It coordinates the production of cytokines including IL-6 G-CSF and GM-CSF and binds to type I and type II OSM receptors forming heterodimers with LIFR/IL6ST and OSMR/IL6ST respectively OSM Protein Human (HEK293 His) is the recombinant human-derived OSM protein expressed by HEK293 with C-His labeled tag