Learn More
Medchemexpress LLC Oncostatin M | 106956-32-5 | MFCD00166731 | >95.0% | 20UG
SDP
Catalog No. 5000177085
Encompass_Preferred
Recombinant mouse Oncostatin M (OSM) expressed in HEK293 cells and supplied lyophilized with a C-terminal His tag. The protein is highly pure (>95% by reducing SDS-PAGE), has an apparent molecular weight of ~33.25 kDa, and shows biological activity in NIH/3T3 proliferation assays (ED50 ≤ 4.975 ng/mL). Intended for cell-based assays and biochemical studies; store frozen and follow reconstitution instructions.
Catalog No. 50-001-77085 Supplier Medchemexpress LLC Supplier No. HYP70080520UG
Recombinant mouse Oncostatin M (OSM) expressed in HEK293 cells and supplied lyophilized with a C-terminal His tag. The protein is highly pure (>95% by reducing SDS-PAGE), has an apparent molecular weight of ~33.25 kDa, and shows biological activity in NIH/3T3 proliferation assays (ED50 ≤ 4.975 ng/mL). Intended for cell-based assays and biochemical studies; store frozen and follow reconstitution instructions.