Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSM™ Natural No-Rinse Floor Cleaner
SCI_ED

Catalog No. S95766
Change view
Click to view available options
Volume (English):
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
S95766 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95766 Supplier OSM™ Supplier No. 81008201219/EMD

missing translation for 'validMainframeMsg'

Cleans gently, leaving a beautiful luster

  • Perfect for cabinets, molding, doors, painted surfaces, and more
  • Use on wood, vinyl, laminate, and tile
  • Penetrates and emulsifies grease, dirt, and grime
  • Highly effective cleaning agents with plant-based surfactants leave no harmful residue
  • Renewable plant-derived formula free of phosphates, glycol ether, and phthalates
  • Safe for use around people and animals
  • Ozone-safe formula disposes down the drain
  • No harsh chemcial fumes or vapor

Specifications

Product Type Floor Cleaner
Form Liquid
Appearance Green
For Use With (Application) For use on tile, laminate, Vinyl, Wood and painted surfaces, safe for cleaning cabinets, molding, paneling and doors
Volume (English) 5 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.