Learn More
Medchemexpress LLC OSM-S-106 | 2650846-59-4 | 99.3% | 306.36 | 5 MG
SDP
Catalog No. 5000392961
Encompass_Preferred
OSM-S-106 is a pro-inhibitor that inhibits PfAsnRS via enzyme-mediated production of an Asn-OSM-S-106 adduct. It inhibits protein translation and activates the amino acid starvation response. This product exhibits selective activity against Plasmodium blood and liver stages and has low intrinsic clearance by human microsomes.
  • Inhibits protein translation
  • Activates the amino acid starvation response
  • Selective activity against Plasmodium blood and liver stages
  • Low intrinsic clearance by human microsomes
Catalog No. 50-003-92961 Supplier Medchemexpress LLC Supplier No. HY1622385MG
OSM-S-106 is a pro-inhibitor that inhibits PfAsnRS via enzyme-mediated production of an Asn-OSM-S-106 adduct. It inhibits protein translation and activates the amino acid starvation response. This product exhibits selective activity against Plasmodium blood and liver stages and has low intrinsic clearance by human microsomes.
  • Inhibits protein translation
  • Activates the amino acid starvation response
  • Selective activity against Plasmodium blood and liver stages
  • Low intrinsic clearance by human microsomes