Learn More
Medchemexpress LLC OSM-S-106 | 2650846-59-4 | 99.3% | 306.36 | 50 MG
SDP
Catalog No. 5000392055
Encompass_Preferred
OSM-S-106 is a pro-inhibitor that works by inhibiting PfAsnRS through an enzyme-mediated adduct formation. It effectively blocks protein translation and triggers an amino acid starvation response. The compound demonstrates selective activity against both Plasmodium blood and liver stages, and exhibits low intrinsic clearance by human microsomes.
  • Inhibits PfAsnRS via an enzyme-mediated adduct.
  • Blocks protein translation.
  • Activates amino acid starvation response.
  • Selectively active against Plasmodium blood and liver stages.
  • Low intrinsic clearance by human microsomes.
Catalog No. 50-003-92055 Supplier Medchemexpress LLC Supplier No. HY16223850MG
OSM-S-106 is a pro-inhibitor that works by inhibiting PfAsnRS through an enzyme-mediated adduct formation. It effectively blocks protein translation and triggers an amino acid starvation response. The compound demonstrates selective activity against both Plasmodium blood and liver stages, and exhibits low intrinsic clearance by human microsomes.
  • Inhibits PfAsnRS via an enzyme-mediated adduct.
  • Blocks protein translation.
  • Activates amino acid starvation response.
  • Selectively active against Plasmodium blood and liver stages.
  • Low intrinsic clearance by human microsomes.