Learn More
Medchemexpress LLC OSM-SMI-10B | 2502294-55-3 | 99.7% | 378.33 | 5 MG
SDP
Catalog No. 5000362268
Encompass_Preferred
OSM-SMI-10B is a derivative of OSM-SMI-10 that significantly reduces OSM-induced STAT3 phosphorylation in cancer cells upon co-incubation with Oncostatin M (OSM). This compound targets p-STAT3 and is involved in JAK/STAT Signaling and Stem Cell/Wnt pathways.
  • Significantly reduces OSM-induced STAT3 phosphorylation
  • Targets p-STAT3
  • Involved in JAK/STAT signaling pathway
  • Involved in stem cell/Wnt pathway
Catalog No. 50-003-62268 Supplier Medchemexpress LLC Supplier No. HY1486925MG
OSM-SMI-10B is a derivative of OSM-SMI-10 that significantly reduces OSM-induced STAT3 phosphorylation in cancer cells upon co-incubation with Oncostatin M (OSM). This compound targets p-STAT3 and is involved in JAK/STAT Signaling and Stem Cell/Wnt pathways.
  • Significantly reduces OSM-induced STAT3 phosphorylation
  • Targets p-STAT3
  • Involved in JAK/STAT signaling pathway
  • Involved in stem cell/Wnt pathway