Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precision Systems Osmette™ Micro-Osmette Osmometer

Catalog No. 128277 Shop All Precision Systems Inc. Products
Change view
Click to view available options
Voltage:
115 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
12-827-7 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-827-7 Supplier Precision Systems Supplier No. 5004
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Automatic digital osmometers measure osmolality in mOsm/kg H2O. Osmette Micro-Osmette Osmometer uses the freezing point depression method to provide a quick, accurate indication of total concentration. Instruments measure freezing points of solutions to 0.001°C, then directly translate them to measure of concentration.

Easy to Use

  • Add sample tube to well, lower operating head to insert wire and thermistor temperature probe into the sample
  • Freezing point is detected by the probe, with accuracy of ±0.001°C
  • Digital display shows results directly in mOsm/kg H2O
  • Entire cycle takes between one-half and one minute per sample
Speedy and convenient
  • Fast cooldown, completely zero-stable meter
  • Reach operating temperature in <15 min. vs. 40 to 60 min. for other cryoscopic devices
  • Eliminate Dewar flasks, ice baths, calculations
  • Generate precise results in just 50 to 70 sec.; process 40 to 60 samples per hour
  • Minimal training; simple two-step calibration takes less than five minutes
  • All models hold calibration, eliminating daily calibration

Micro-Osmette, Model 5004

  • Automatic Operation
  • Features electronic cooling instead of a bath
  • Food and beverage industries, chemical manufacturing, pharmaceutical labs, research and quality control
  • Clinical applications include diagnosis of renal activity, disease electrolyte imbalance, hyper- and hyponatremia, measuring osmolality of body fluids

  1. Reproducibility, 1 S.D. (one standard deviation).
  2. With liquid bath.
  3. Also available: Cryette™ Cryoscopes. Model 5006 (Cat. No. 12-672) determines added water in milk; Models 5008 and 5009 (Cat. Nos. 12-672-1 and 12-672-2) determine molecular weight of solutions such as petroleum fractions and core samples. For information, contact your Fisher Scientific Customer Service Representative.

Specifications

Display Type Digital
Frequency 50/60 Hz
Voltage 115 V
Cooling Agent Thermoelectric Cooling Block
Quantity Each
Product Type Micro Osmometer
Model 5004
For Use With (Application) Food and beverage industries, chemical manufacturing, pharmaceutical labs, research and quality control, clinical applications include diagnosis of renal activity, disease electrolyte imbalance, hyper- and hyponatremia, measuring osmolality of body fluids
Height (English) 10 in.
Height (Metric) 25 cm
Length (English) 15 in.
Measurement Time <1 min.
Length (Metric) 38 cm
Operation Automatic
Precision <1000mOsm/kg H2O: ±2mOsm/kg H2O, >1000mOsm/kg H2O: ±0.5%CV at 300mOsm/kg H2O: <1%
Range 0 to 3000 mOsm/kg H2O
Sample Size 50 μL
Width (Metric) 13 cm
Width (English) 5 in.
Cool-Down Time ∼3 min.
Dimensions (L x W x H) 15 x 5 x 10 in. (38 x 13 x 25 cm)
Electrical Requirements 115 V 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.