Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precision Systems Bath Fluid for Osmette™ Osmometers

Catalog No. 1282722 Shop All Precision Systems Inc. Products
Change view
Click to view available options
Volume (Metric):
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12-827-22 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-827-22 Supplier Precision Systems Supplier No. 2100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Osmette A Osmometer, Osmette XL Osmometer.

  • Quantity: 250mL bottle

Specifications

Product Type Osmometer Bath Fluid
For Use With (Equipment) 12-8275, -3 and -33
Volume (English) 8.45 oz.
Volume (Metric) 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.