Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precision Systems Osmette™ Osmometer Reusable Sample Tubes, 2 mL

Catalog No. 128272 Shop All Precision Systems Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Freezing Point Osmometer Model 5002 Osmette A
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
12-827-2 Freezing Point Osmometer Model 5002 Osmette A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-827-2 Supplier Precision Systems Supplier No. 2021
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glass sample tubes for Models 5002 Osmette A and 5007 Osmette XL

  • For Precision Systems Model Numbers: 5002 and 5007

Specifications

Product Type Sample Tube
For Use With (Equipment) Freezing Point Osmometer Model 5002 Osmette A
Disposable Single-use
Volume (English) 0.06 oz.
Volume (Metric) 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.