Learn More
Precision Systems Osmette™ Sample Tubes, 30 μL
Catalog No. 01184160 Shop All Precision Systems Inc. Products
Change view
Click to view available options
Volume (Metric):
30 μL
Catalog No. Volume (Metric)
01-184-160 30 μL
1 options
This item is not returnable. View return policy
Catalog No. 01-184-160 Supplier Precision Systems Supplier No. 2038
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sample tubes for Models 2430 Multi-Osmette and 6002, and 6003 Touch-Micro Osmette Osmometers
  • For Precision Systems Model Numbers: 2430, 6002, and 6003 Osmometers
Specifications
Product Type Sample Tube
For Use With (Equipment) Osmometers and Cryoscopes
Disposable Single-use
Volume (Metric) 30 μL