Learn More
TARGETMOL CHEMICALS INC OSMI-1 10MG
SDP
Catalog No. 502577339
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OSMI-1 inhibits protein O-linked N-acetylglucosamine (O-GlcNAcylation) in several mammalian cell lines without qualitatively altering cell surface N- or O-linked glycans. OSMI-1 is a cell-permeable O-GlcNAc transferase (OGT) inhibitor (IC50 2.7 uM). purity: 99%
Catalog No. 50-257-7339 Supplier TARGETMOL CHEMICALS INC Supplier No. T1640910MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OSMI-1 inhibits protein O-linked N-acetylglucosamine (O-GlcNAcylation) in several mammalian cell lines without qualitatively altering cell surface N- or O-linked glycans. OSMI-1 is a cell-permeable O-GlcNAc transferase (OGT) inhibitor (IC50 2.7 uM). purity: 99%