Learn More
Selleck Chemical LLC OSMI-4 5mg
SDP
Supplier:  Selleck Chemical LLC S89105MG
Encompass_Preferred
OSMI-4 is an inhibitor of O-linked N-acetylglucosamine transferase (O-GlcNAc transferase, OGT) with EC50 of 3 ?M in cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3429