Learn More
Medchemexpress LLC HY-114361 5mg Medchemexpress, OSMI-4 CAS:2260791-14-6 Purity:>98%
SDP
Catalog No. 501932480
Encompass_Preferred
Medchemexpress, HY-114361 5mg OSMI-4 CAS:2260791-14-6 OSMI-4 is a low nanomolar O-GlcNAc transferase (OGT) inhibitor, with an EC 50 of 3 μM in cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2480 Supplier Medchemexpress LLC Supplier No. HY1143615MG
Medchemexpress, HY-114361 5mg OSMI-4 CAS:2260791-14-6 OSMI-4 is a low nanomolar O-GlcNAc transferase (OGT) inhibitor, with an EC 50 of 3 μM in cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.