Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osmic Acid Anhydride (Crystalline/Laboratory), Fisher Chemical™

Catalog No. A23514 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
0.25 g
1 g
Packaging:
Ampule
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A235-14 0.25 g Ampule
A235-1 1 g Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A235-14

missing translation for 'validMainframeMsg'

Safety seal, prescored neck, easy-to-snap-open ampules

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Boiling Point 130°C
Melting Point 39.5°C
Color White
Physical Form Solid
Quantity 0.25 g
Formula Weight 254.23
Grade Laboratory
Identification Pass Test
Packaging Ampule
Vapor Pressure 7mmHg at 20°C
Density 4.9g/cm³
Chemical Name or Material Osmic Acid Anhydride
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 6.1, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.