Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osmium 99.9%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
7440-04-2
Os
190.23
MFCD00011147
SYQBFIAQOQZEGI-UHFFFAOYSA-N
ion,metallic,osmium, elemental,unii-2e7m255opy,os4,osmio,hydrido,hydride,atom,powder
23937
CHEBI:30687
osmium
[Os]
7440-04-2
Os
190.23
MFCD00011147
SYQBFIAQOQZEGI-UHFFFAOYSA-N
ion,metallic,osmium, elemental,unii-2e7m255opy,os4,osmio,hydrido,hydride,atom,powder
23937
CHEBI:30687
osmium
[Os]
for ultra trace analysis
Quantity 5g
Assay Percent Range 99.90%
Linear Formula Os
Merck Index 15, 6988
Solubility Information Solubility in water: insoluble.
Packaging Glass bottle
Formula Weight 190.2
Physical Form Powder
Percent Purity 99.9%
Grade Trace Metal Basis
Total Trace Metal Impurities 0.1% max.
Color Black
Melting Point 3045.0°C
Boiling Point 5027.0°C
Chemical Name or Material Osmium
Show More Show Less

GHS H Statement:
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning