Learn More
Strem, An Ascensus Company Osmium carbonyl, 99% 2g
SDP
Catalog No. 502034198
Encompass
CAS# 15696-40-9. 2g. Osmium carbonyl, 99%. MFCD00011149. Molecular Weight: 906.73. Molecular Formula: Os3(CO)12. Color/form: yellow xtl. Strem# 76-2000. http://www.strem.com/catalog/v/76-2000/
Catalog No. 50-203-4198 Supplier Strem, An Ascensus Company Supplier No. 7620002G
Add to Cart
Add to Cart
Request a Quote
CAS# 15696-40-9. 2g. Osmium carbonyl, 99%. MFCD00011149. Molecular Weight: 906.73. Molecular Formula: Os3(CO)12. Color/form: yellow xtl. Strem# 76-2000. http://www.strem.com/catalog/v/76-2000/