Learn More
Electron Microscopy Sciences EMS Osmium Tetroxide 2% Aqueous Solution 10 ML
SDP
Supplier:  Electron Microscopy Sciences 19192
Encompass_Preferred
To facilitate your work, we now have available a complete line of Osmium Tetroxide Aqueous Solutions with various concentrations. Sealed under dry nitrogen. Prepared in our plant daily. Exact concentrations and volume in pre-scored amber-ampoules.
Catalog No. 50-332-21