Learn More
Electron Microscopy Sciences EMS Osmium Tetroxide, 2% Aqueous Solution 2 ML
SDP
Catalog No. 5033218
Encompass_Preferred
To facilitate your work, we now have available a complete line of Osmium Tetroxide Aqueous Solutions with various concentrations. Sealed under dry nitrogen. Prepared in our plant daily. Exact concentrations and volume in pre-scored amber-ampoules.
Catalog No. 50-332-18 Supplier Electron Microscopy Sciences Supplier No. 19152
To facilitate your work, we now have available a complete line of Osmium Tetroxide Aqueous Solutions with various concentrations. Sealed under dry nitrogen. Prepared in our plant daily. Exact concentrations and volume in pre-scored amber-ampoules.