Learn More
Ted Pella Inc Osmium Tetroxide, 20816-12-0, MFCD00011150, 4% aqueous, 10 x 5ml
SDP
Catalog No. NC1037266
Encompass
Linear Formula: OsO4, Molecular Weight: 254.23, Synonym: ‘Osmic acid’, Osmium(VIII)-oxide, BP: 130 °C(lit.), MP: 39.5-41 °C.
Catalog No. NC1037266 Supplier Ted Pella Inc Supplier No. 18463
Linear Formula: OsO4, Molecular Weight: 254.23, Synonym: ‘Osmic acid’, Osmium(VIII)-oxide, BP: 130 °C(lit.), MP: 39.5-41 °C.