Learn More
Electron Microscopy Sciences EMS Osmium Tetroxide 4% Aqueous Solution 10 ML
SDP
Catalog No. 5033210
Encompass_Preferred
To facilitate your work, we now have available a complete line of Osmium Tetroxide Aqueous Solutions with various concentrations. Sealed under dry nitrogen. Prepared in our plant daily. Exact concentrations and volume in pre-scored amber-ampoules.
Catalog No. 50-332-10 Supplier Electron Microscopy Sciences Supplier No. 19180
To facilitate your work, we now have available a complete line of Osmium Tetroxide Aqueous Solutions with various concentrations. Sealed under dry nitrogen. Prepared in our plant daily. Exact concentrations and volume in pre-scored amber-ampoules.