Learn More
Crescent Chemical Co Inc OSMIUM TETROXIDE 4% SOLUTION
SDP
Catalog No. 50247967
Encompass_Preferred
Osmium tetroxide 4 % solution for electron microscopy 40 mg/ml in water. Size - 5x10ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 31253.04
Catalog No. 50-247-967 Supplier Crescent Chemical Co Inc Supplier No. SE31253.04
Osmium tetroxide 4 % solution for electron microscopy 40 mg/ml in water. Size - 5x10ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 31253.04