Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osmium tetroxide, 99.9+%, (trace metal basis)

Catalog No. AC191185000
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g

CAS: 20816-12-0 | O4Os | 254.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Boiling Point 130.0°C
Melting Point 40.0°C to 42.0°C
Color Colorless to Yellow
Physical Form Crystals
Quantity 500 mg
Assay Percent Range 99.9+%
Linear Formula OsO4
Fieser 01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236
Merck Index 15, 6990
Solubility Information Passes test
Formula Weight 254.2
Percent Purity ≥99.9%
Grade Trace Metal Basis
Packaging Ampoule
Chemical Name or Material Osmium tetroxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Fatal if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
Rinse mouth.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/f
EINECSNumber 244-058-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.