Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osmium Tetroxide, ACS Reagent, ≥98%, Solstice

Catalog No. 60046847 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-847 250 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-046-847 Supplier Solstice Supplier No. 419494250MG

missing translation for 'validMainframeMsg'

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Boiling Point 130°C
Melting Point 39.5°C to 41°C
Physical Form Solid
Quantity 250 mg
Linear Formula OsO4
UN Number UN2471
Solubility Information Slightly soluble in water
Formula Weight 254.23g/mol
Percent Purity ≥98%
Odor Unpleasant
Packaging Glass Bottle
Vapor Pressure 7mmHg (20°C)
Density Vapor Density: 8.8 (vs air)
Chemical Name or Material Osmium tetroxide
Show More Show Less
Hazard Statement P260-P264-P280-P284-P301 + P310-P302 + P350
DOTInformation 6.1/PGI
RTECSNumber 244-058-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.