Learn More
Electron Microscopy Sciences Osmium Tetroxide, 20816-12-0, MFCD00011150, 1 g
SDP
Catalog No. 5027578
Encompass_Preferred
Formula: OsO4, MW: 254.20, Crystalline, Highest Purity, 99.95%, Melting Range: 39.5-41.0°C, Boiling Point: 129.7°C.
Catalog No. 50-275-78 Supplier Electron Microscopy Sciences Supplier No. 19100
Formula: OsO4, MW: 254.20, Crystalline, Highest Purity, 99.95%, Melting Range: 39.5-41.0°C, Boiling Point: 129.7°C.