Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences EMS Osmium Tetroxide, Crystalline 0.1 GR
SDP

Catalog No. 5027580
Encompass_Preferred

(Osmic Acid Anhydride)
CAS #20816-12-0 OsO₄ F.W. 254.20
Solubility: 5.07% in Cold Water; 6.5% in Hot Water; Moderately sol. (7 g/100 mL), 25.0% in CCl4
Melting point: 39.5-41.0°C
Boiling point: 129.7°C

Crystalline, highest purity (99.95%), osmic acid anhydride. Small size (2ml), light-gauge, pre-scored amber glass ampoules. Ampoules containing osmium tetroxide crystal are pre-cleaned and heat sealed in plastic bags to assure a contaminant-free solution. Ampoules are also free of pressure-sensitive labels and, therefore, do not require cleaning prior to solution preparation.

Catalog No. 50-275-80 Supplier Electron Microscopy Sciences Supplier No. 19134
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Osmic Acid Anhydride)
CAS #20816-12-0 OsO₄ F.W. 254.20
Solubility: 5.07% in Cold Water; 6.5% in Hot Water; Moderately sol. (7 g/100 mL), 25.0% in CCl4
Melting point: 39.5-41.0°C
Boiling point: 129.7°C

Crystalline, highest purity (99.95%), osmic acid anhydride. Small size (2ml), light-gauge, pre-scored amber glass ampoules. Ampoules containing osmium tetroxide crystal are pre-cleaned and heat sealed in plastic bags to assure a contaminant-free solution. Ampoules are also free of pressure-sensitive labels and, therefore, do not require cleaning prior to solution preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.