Learn More
Sigma Aldrich Fine Chemicals Biosciences Osmium tetroxide ReagentPl1G
Catalog No. 502897358
Encompass
Osmium tetroxide (OsO4) is widely employed as a catalyst in various organic reactions. Stereochemical aspects of the OsO4 assisted oxidation of allylic alcohols and their derivatives have been reported. Its reaction with olefins has been investigated. It participates as a catalyst for oxidizing fluoroquinolones via reaction with potassium hexacyanoferrate(III) in alkaline medium. Kinetics of this oxidation reaction was studied.
Catalog No. 50-289-7358 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2010301G
Osmium tetroxide (OsO4) is widely employed as a catalyst in various organic reactions. Stereochemical aspects of the OsO4 assisted oxidation of allylic alcohols and their derivatives have been reported. Its reaction with olefins has been investigated. It participates as a catalyst for oxidizing fluoroquinolones via reaction with potassium hexacyanoferrate(III) in alkaline medium. Kinetics of this oxidation reaction was studied.