Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osmium Tetroxide Solution, Solstice

Catalog No. 60045177 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-177 5 mL Glass Bottle
60-045-176 25 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-177 Supplier Solstice Supplier No. 2088685ML

2.5 wt. % in tert-butanol

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Physical Form Liquid
Quantity 5 mL
Linear Formula OsO4
UN Number UN3286
Flash Point 4°C (39.2°F)
Formula Weight 254.23g/mol
Packaging Glass Bottle
Vapor Pressure 6.76psi (55°C), 5.42psi (20°C)
Density 1.0g/mL at 20°C, 0.811g/mL at 25°C
Chemical Name or Material Osmium Tetroxide Solution, Contains tert-butyl hydroperoxide as stabilizer
Hazard Statement P210-P280-P304 + P340 + P310-P305 + P351 + P338 + P310-P370 + P378-P403 + P235
Precautionary Statement H225-H301-H312-H315-H317-H318-H330-H334-H335-H336-H341
DOTInformation 8(6.1)(3)/PGII
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.