Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osmium Tetroxide Solution, Sigma-Aldrich

Catalog No. 01258863 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-863 5 mL
01-258-988 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-863 Supplier MilliporeSigma Supelco Supplier No. 756325ML
Add to Cart
Add to Cart

Due to the hazardous nature of osmium tetroxide, it is recommended to use an ampule opener when using this chemical.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Osmium Tetroxide
CAS 20816-12-0
Concentration 4% in H2O
Molecular Weight (g/mol) 254.23 g/mol
Color Colorless to Yellow
pH 6 to 8
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.