Learn More
Strem, An Ascensus Company CAS# 20816-12-0. 10ml. Osmium(VIII) oxide, 4% in water. MFCD00011150
SDP
Catalog No. 5090113091
Encompass
CAS# 20816-12-0. 10ml. Osmium(VIII) oxide, 4% in water. MFCD00011150. Molecular Weight: . Molecular Formula: OsO4. Color/form: colorless liq. Strem# 76-2952. http://www.strem.com/catalog/v/76-2952/
Catalog No. 50-901-13091 Supplier Strem, An Ascensus Company Supplier No. 76295210ML
CAS# 20816-12-0. 10ml. Osmium(VIII) oxide, 4% in water. MFCD00011150. Molecular Weight: . Molecular Formula: OsO4. Color/form: colorless liq. Strem# 76-2952. http://www.strem.com/catalog/v/76-2952/