Learn More
Strem, An Ascensus Company CAS# 20816-12-0. 500mg. Osmium(VIII) oxide (99.95+%-Os). MFCD00011150
SDP
Catalog No. 5090113086
Encompass
CAS# 20816-12-0. 500mg. Osmium(VIII) oxide (99.95+%-Os). MFCD00011150. Molecular Weight: 254.20. Molecular Formula: OsO4. Color/form: pale yellow xtl. Strem# 76-2950. http://www.strem.com/catalog/v/76-2950/
Catalog No. 50-901-13086 Supplier Strem, An Ascensus Company Supplier No. 762950500MG
CAS# 20816-12-0. 500mg. Osmium(VIII) oxide (99.95+%-Os). MFCD00011150. Molecular Weight: 254.20. Molecular Formula: OsO4. Color/form: pale yellow xtl. Strem# 76-2950. http://www.strem.com/catalog/v/76-2950/