Learn More
NewPath Learning™ Osmosis and Diffusion Student Learning Guide with Online Lesson
SCI_ED
Catalog No. S436009 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
Catalog No. Grade
S436009 6 to 10
1 options
This item is not returnable. View return policy
Catalog No. S436009 Supplier NewPath Learning™ Supplier No. 146743
Add to Cart
Add to Cart
This item is not returnable. View return policy
An ideal blended learning solution for use in the classroom or for remote learning/home study.
  • Colorfully-illustrated Student Learning Guide provides 36 pages featuring Self-Directed Readings Easy-to-Follow Illustrated Explanations Guiding Questions a Lab Investigation Key Vocabulary Review and Assessments.
  • Also includes one year access to a corresponding Multimedia Lesson for a teacher and all their students Each lesson features Animated Narrated Tutorials Interactive Exercises and Activities Virtual Lab Investigations Illustrated Narrated Glossary Visual Resource Library and an Inquiry-Based Student Activity Guide.
  • Developed under sponsorship of the National Institutes of Health (NIH) the lessons are perfect for individual student instruction on any computer or tablet as well as classroom presentation on any Interactive Whiteboard.
  • Covers Comparing Plant and Animal Cells Cell Membrane and Cell Transport Diffusion Diffusion in the Lungs Osmosis The Diffusion of Water Passive Transport Active Transport and Osmosis in Plant and Animal Cells.
Specifications
Grade Level Intermediate - Middle School, High School
Grade 6 to 10
Category Life Science
Includes Student Learning Guide, Card with Link and Code to Online Multimedia Lesson