Learn More
Aviva Systems Biology Corp OSMR Antibody - middle region (ARP46396_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46396_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OSMR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OSMR. Alternate names: MGC150626, MGC150627, MGC75127, OSMRB, PLCA1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rabbit
Catalog No. 50-014-59319