Learn More
Medchemexpress LLC OSMR beta Human 71 10ug
SDP
Catalog No. 5000182144
Encompass_Preferred
The OSMR beta protein is critical in immune responses by binding to IL12RB1 to form the interleukin 23 receptor with IL-23R It binds to IL23 and activates T cells NK cells and potentially macrophages/myeloid cells through Jak-Stat signaling OSMR beta Protein Human (712a a HEK293 His) is the recombinant human-derived OSMR beta protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-82144 Supplier Medchemexpress LLC Supplier No. HYP7250010UG
The OSMR beta protein is critical in immune responses by binding to IL12RB1 to form the interleukin 23 receptor with IL-23R It binds to IL23 and activates T cells NK cells and potentially macrophages/myeloid cells through Jak-Stat signaling OSMR beta Protein Human (712a a HEK293 His) is the recombinant human-derived OSMR beta protein expressed by HEK293 with C-6 His labeled tag