Learn More
Medchemexpress LLC OSMR beta Mouse HE 5ug | 5ug
SDP
Catalog No. 5000193912
Encompass_Preferred
The OSMR beta Protein in partnership with IL31RA constitutes the IL31 receptor binding IL31 to activate STAT3 and potentially STAT1 and STAT5 OSMR beta also transduces OSM-specific signaling It forms a heterodimer with IL6ST creating the type II OSM receptor and another heterodimer with IL31RA for the IL31 receptor OSMR beta Protein Mouse (HEK293 His) is the recombinant mouse-derived OSMR beta protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-93912 Supplier Medchemexpress LLC Supplier No. HYP783335UG
The OSMR beta Protein in partnership with IL31RA constitutes the IL31 receptor binding IL31 to activate STAT3 and potentially STAT1 and STAT5 OSMR beta also transduces OSM-specific signaling It forms a heterodimer with IL6ST creating the type II OSM receptor and another heterodimer with IL31RA for the IL31 receptor OSMR beta Protein Mouse (HEK293 His) is the recombinant mouse-derived OSMR beta protein expressed by HEK293 with C-His labeled tag