Learn More
Eurofins Discoverx PathHunter OSMRb/IL31R Bioassay Kit, 10-Plate. (93-1002Y3-00084)
Catalog No. 502001808
Encompass
The PathHunter OSMRb/IL31R Dimerization Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. IL-31) induced dimerization of OSMRb/IL31R receptors. This product uses EFC technology. The cell line included overexpresses OSMRb target protein fused to PK alongside IL31RA target protein fused to EA. Upon ligand-induced dimerization of OSMRb/IL31R, detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-1808 Supplier Eurofins Discoverx Supplier No. 931002Y300084
Add to Cart
Add to Cart
Request a Quote
The PathHunter OSMRb/IL31R Dimerization Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. IL-31) induced dimerization of OSMRb/IL31R receptors. This product uses EFC technology. The cell line included overexpresses OSMRb target protein fused to PK alongside IL31RA target protein fused to EA. Upon ligand-induced dimerization of OSMRb/IL31R, detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.