Learn More
Cayman Chemical Osntnthydrate 10mg
SDP
Catalog No. 502948276
Encompass_Preferred
An NK3 receptor antagonist (Ki = 0.21 nM in CHO cells expressing the human receptor); selective for NK3 over NK1 and NK2 receptors (Kis = >100 and 0.02 µM, respectively); inhibits neurokinin B-induced contractions and acetylcholine release in isolated guinea pig ileal strips; inhibits ovalbumin-induced increases in BALF neutrophil, eosinophil, and lymphocyte infiltration in an ovalbumin-sensitized mouse model of allergic asthma; has anxiolytic-like and antidepressant-like activities in gerbils at 5 and 10 mg/kg
Catalog No. 50-294-8276 Supplier Cayman Chemical Supplier No. 3341710 MG
An NK3 receptor antagonist (Ki = 0.21 nM in CHO cells expressing the human receptor); selective for NK3 over NK1 and NK2 receptors (Kis = >100 and 0.02 µM, respectively); inhibits neurokinin B-induced contractions and acetylcholine release in isolated guinea pig ileal strips; inhibits ovalbumin-induced increases in BALF neutrophil, eosinophil, and lymphocyte infiltration in an ovalbumin-sensitized mouse model of allergic asthma; has anxiolytic-like and antidepressant-like activities in gerbils at 5 and 10 mg/kg