Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics™ OSOM™ BVBLUE™ Test

Catalog No. 21150120 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Type:
BVBLUE Test Kit
Control Kit
Transport Swabs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
21-150-120 Transport Swabs
22-071-073 BVBLUE Test Kit
22-071-074 Control Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 21-150-120 Supplier SEKISUI Diagnostics™ Supplier No. 186
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use the OSOM™ BVBLUE™ Test to diagnose bacterial vaginosis (BV) in just ten minutes for faster treatment and education in the same office visit.

  • Test for the four major pathogens that cause BV in just one test - reacts with sialidase produced by Gardnerella vaginalis, Bacteroides spp., Prevotella, and Mobiluncus spp.
  • Simple-to-use - get easy to read results in four easy steps and no need for additional equipment
  • Decrease the chance of human error with minimal hands-on requirements
  • Quicker results - read results in just 10 minutes
  • Easy-to-read, instant color change for accurate results
  • Get more patient tests per kit - exclusive QC Inside™ provides two extra tests at no additional charge to run external quality control testing
  • Made in the USA
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
CPT Code 87905QW
Clia Complexity Waived
Detectable Analytes Sialidase
Sample Type Vaginal Swab
Sensitivity 92.8%
Type Transport Swabs
Includes 30 swabs
Packaging Type Test Kit
Specificity 0.98
Test Time 10 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.