Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics™ OSOM™ BVBLUE™ Test
GSA_VA

Catalog No. 22071073 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Type:
BVBLUE Test Kit
Control Kit
Transport Swabs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-071-073 BVBLUE Test Kit
21-150-120 Transport Swabs
22-071-074 Control Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-071-073 Supplier SEKISUI Diagnostics™ Supplier No. 183
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use the OSOM™ BVBLUE™ Test to diagnose bacterial vaginosis (BV) in just ten minutes for faster treatment and education in the same office visit.

  • Test for the four major pathogens that cause BV in just one test - reacts with sialidase produced by Gardnerella vaginalis, Bacteroides spp., Prevotella, and Mobiluncus spp.
  • Simple-to-use - get easy to read results in four easy steps and no need for additional equipment
  • Decrease the chance of human error with minimal hands-on requirements
  • Quicker results - read results in just 10 minutes
  • Easy-to-read, instant color change for accurate results
  • Get more patient tests per kit - exclusive QC Inside™ provides two extra tests at no additional charge to run external quality control testing
  • Made in the USA

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Refrigerate
CPT Code 87905QW
Clia Complexity Waived
Detectable Analytes Sialidase
Format Kit
Sample Type Vaginal Swab
Sensitivity 92.8%
Type BVBLUE Test Kit
For Use With (Equipment) None
Includes 25 test vessels, 1 bottle of developer solution, sterile swabs, 1 instruction for use (IFU)
Packaging Type Test Kit
Specificity 98%
Test Time 10 min.
Analysis Time 10 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.