Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics™ OSOM™ COVID-19 Antigen Home Test

Catalog No. 22321138 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
SEKISUI Diagnostics™ COVID-19 Over-the-Counter Test
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-321-138 SEKISUI Diagnostics™ COVID-19 Over-the-Counter Test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-321-138 Supplier SEKISUI Diagnostics™ Supplier No. 106740
This item is not returnable. View return policy

OSOM™ COVID-19 Antigen home test reliably detects SARS-CoV-2 in only 15 minutes, without a doctor's visit or prescription.

Trust your results - The same professional-grade technology that doctors have trusted and used for years

  • Easy-to-use - Can be performed in the privacy and comfort of your own home without the need for a doctor's visit or prescription
  • Get back to normal, faster - Everything you need to test for COVID-19 in only 15 minutes
  • Get it when you need it - Proudly made in the USA and backed by a trustworthy supply chain
  • Exceptional support - Responsive support teams and in-depth online product information so you are not left without answers
TRUSTED_SUSTAINABILITY

Specifications

Accuracy 84.4% (PPA) and 100% (NPA)
Buffer Ready-to-Use (RTU)
Content And Storage Room temperature
CE Marker No
Clia Complexity Waived (EUA)
Detectable Analytes SARS-CoV-2
Detection Method Chromatographic
Format Cassette
Sample Type Nasal
Sensitivity 84.40%
Type Over-the-Counter COVID-19 Test
Disposable Single-use
For Use With (Application) Laterial Flow Immunoassay
Sterility Sterile
Analysis Time 15 minutes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.