Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sekisui Diagnostics™ OSOM™ COVID-19 Antigen Rapid Test

Catalog No. 22321136 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
COVID-19 Antigen Rapid Test; Clia Complexity: Waived (EUA)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-321-136 COVID-19 Antigen Rapid Test; Clia Complexity: Waived (EUA)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-321-136 Supplier Sekisui Diagnostics™ Supplier No. 106640
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Reliably detect SARS-CoV-2 in only 15 minutes at the point-of-care.

  • High positive percent agreement and negative percent agreement to help you reduce the risk of misdiagnosis and inappropriate treatment
  • Easy to use: Can be performed in a CLIA-waived setting and comes with everything you need to run a test without the need for specialized equipment
  • Get results to your patient in 15 minutes, with the ability to run multiple tests simultaneously
  • Simple sample collection: Direct Mid-Turbinate nasal swab to help you improve patient comfort and satisfaction
  • Proudly made in the USA and backed by a trustworthy supply chain so you can avoid delays from international shipments
  • Experienced support teams and online training modules to help streamline your testing implementation and staff training
TRUSTED_SUSTAINABILITY

Specifications

CE Marker No
Clia Complexity Waived (EUA)
Detectable Analytes SARS-CoV-2
Detection Method Antigen
Format Cassette
Sample Type Nasal Swab
Sensitivity High
Type Rapid Test
Disposable Single-use
For Use With (Application) Laterial Flow Immunoassay
Sterility Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.