Description
- Lateral flow immunochromatographic assay intended for the qualitative detection and differentiation of influenza A, and influenza B nucleoprotein antigens and SARS-CoV-2 nucleocapsid antigen directly in anterior nasal swab specimens from individuals with signs and symptoms of respiratory tract infection
- QC Inside: Two extra test devices have been included for external QC testing
Specifications
Specifications
| CE Marker | No |
| Clia Complexity | Waived |
| Detectable Analytes | SARS-CoV-2, Influenza A, Influenza B |
| Type | Combo Test |
| Disposable | Single-use |
| Form | Swab |
For prescription use only.