Learn More
Sekisui Diagnostics OSOM™ COVID-19 Test
Catalog No. 22321107 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Detectable Analytes:
SARS-CoV-2 Nucleocapsid Protein Antigen
Catalog No. Detectable Analytes
22-321-107 SARS-CoV-2 Nucleocapsid Protein Antigen
1 options
Catalog No. 22-321-107 Supplier Sekisui Diagnostics Supplier No. 1092
Add to Cart
Add to Cart
Reliable testing - Test patients of all ages within 4 days of symptom onset to avoid delaying treatment.
  • A lateral flow immunochromatographic assay for the rapid, qualitative detection of SARS-CoV-2 nucleoprotein antigens directly in anterior nasal swab specimens from individuals with signs and symptoms of upper respiratory infection (symptomatic) when testing is initiated within 4 days of symptom onset
  • Rapid-deliver test results to your patients in only 15 minutes, at the point-of-care
  • Easy to Use: A simple, CLIA-waived procedure that does not require disposable pipettes for dispensing
  • Cost Efficient: Unique QC Inside™ feature that provides you with two additional test for external QC testing
  • Exceptional Support: Experienced support teams and online training modules to help streamline your testing implementation
  • Reagents and Materials Provided:
    • 27* Test devices in sealed aluminum foil pouch with desiccant
    • 27* Reagent tubes with extraction buffer (0.3 mL)
    • 25 Sample collection swabs
    • 1 Positive control swab
    • 1 Negative control swab
    • 1 Instructions for Use (IFU)
    • 1 Quick Reference Guide (QRG)
  • Note: Two extra test devices have been included for external QC testing
TRUSTED_SUSTAINABILITY
Specifications
CE Marker No
Clia Complexity Waived
Detectable Analytes SARS-CoV-2 Nucleocapsid Protein Antigen
Type COVID Test
Disposable Single-use
Sterility Non-sterile
Form Swab
For prescription use only.