Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics™ OSOM™ iFOB Test

Catalog No. 22321112 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Type:
Control Kit
Patient Sample Mailers
Rapid Test Kits
Stool Collection Tubes
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-321-112 Control Kit
22-321-109 Rapid Test Kits
22-321-110 Patient Sample Mailers
22-321-111 Stool Collection Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-321-112 Supplier SEKISUI Diagnostics™ Supplier No. 1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

OSOM iFOB Test (Cat. No. 22-321-109)
  • 25-OSOM iFOB Test device in a sealed foil pouch
  • 25 Stool collection tube, with sampler, containing extraction buffer solution (PBS)
  • 25 Collection paper
  • 25 Mailing Envelope
  • Instructions for use
  • 25 Patient instructions for use
  • 25 Biohazard labeled bags with absorbent pads
  • Two extra test devices for external QC testing

OSOM iFOBT Control Kit (Cat. No. 22-321-112)
1mL OSOM iFOBT Control Positive Control and 1mL OSOM iFOBT Control Negative Control

OSOM iFOBT Patient Mailers (Cat. No. 22-321-110)

25 Mailers

OSOM iFOBT Stool Collection Tubes (Cat. No. 22-321-111)
25 Collection tubes

OSOM™ iFOB Test is an easy-to-use rapid test that provides quick, accurate results for detection of fecal occult blood or human hemoglobin for simpler diagnosis of gastrointestinal disorders.

The OSOM™ iFOB Test is a rapid qualitative test for the immunochemical detection of fecal occult blood/human hemoglobin (hHb) in human fecal specimens as an aid in the diagnosis of gastrointestinal disorders such as: diverticulitis, colitis, polyps, and colorectal cancer. Convenient take home kit, patients can collect in the privacy of their own home to increase compliance in screening.

  • Aids in prevention and helps reduce the risk of developing more serious ulcers and other health issues
  • Lower healthcare costs - early diagnosis leads to the reduction of cancer treatment expenses, hospital stays, and long-term care costs
  • Improved patient compliance - no dietary or drug restrictions, with an easy-to-use at home sample collection kit and procedure
  • Quick turnaround - obtain results in 5 to 10 minutes with less than 1 minute of hands-on time
  • Trust your results - highly specific to human hemoglobin with analytical sensitivity of 50 ng hHb/mL buffer or 50 μg hHb/g feces
  • QC Inside™ - two extra tests included at no additional charge to run external quality control testing for more confidence in your results

Recommended Storage

Store the kit or kit contents 2° to 30°C until use. Do not freeze.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 30°C (35.6°F to 86°F)
Clia Complexity Not Applicable
Detectable Analytes Hemoglobin
Sensitivity 50 ng hHb/mL Buffer or 50 μg hHb/g Feces
Type Control Kit
For Use With (Application) Fecal Occult Blood Detection
Includes 1 mL OSOM iFOBT Control Positive Control and 1 mL OSOM iFOBT Control Negative Control
Specificity Human hemoglobin
Test Time 5 min.
Analysis Time 5 min.

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.