Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sekisui Diagnostics OSOM RSV Test

Catalog No. 21403210 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
CE Marker:
No
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. CE Marker
21-403-210 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-403-210 Supplier Sekisui Diagnostics Supplier No. 1091
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapid immuno chromatographic assay for the qualitative detection of respiratory syncytial virus (RSV) nucleoprotein antigen in anterior nasal swab specimens from patients with signs and symptoms of respiratory infections.

  • The OSOM™ RSV Test is a rapid immunochromatographic assay for the qualitative detection of respiratory syncytial virus (RSV) nucleoprotein antigen in anterior nasal swab specimens from patients with signs and symptoms of respiratory infections
  • This test is intended for in vitro diagnostic use to aid in the diagnosis of RSV infections in infants and pediatric patients aged 6 months to 6 years old, and adults over 60 years of age
  • Test results are interpreted at 15 minutes visually
  • REAGENTS AND MATERIALS Provided
  • 27* - Test devices in sealed aluminum foil pouch with desiccant
  • 27* - Reagent tubes prefilled with extraction buffer (350 μL) and dropper tips
  • 25 - Sample collection swabs (Anterior Nasal)
  • 1 - Positive control swab (noninfectious RSV antigen)
  • 1 - Negative control swab (noninfectious nasal clinical matrix)
  • 1 - Instructions for Use (IFU)
  • 1 - Quick Reference Guide (QRG)
  • NOTE: Two extra test devices have been included for external QC testing.
TRUSTED_SUSTAINABILITY

Specifications

CE Marker No
Clia Complexity Waived
Type Anterior Nasal Swab
For Use With (Application) RSV
Form Swab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.