Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics™ OSOM™ Ultra hCG Combo Rapid Test

Catalog No. 22321113 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Test Time:
3 to 5 min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Test Time
22-321-113 3 to 5 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-321-113 Supplier SEKISUI Diagnostics™ Supplier No. 1004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

OSOM™ Ultra hCG Combo Rapid Test is ideal for facilities testing women beyond the first trimester of pregnancy—ability to detect intact hCG and beta (β) subunit of hCG in urine and serum.

Assay has the ability to detect intact hCG as well as hCG beta (β) subunits and some hCG fragments. The test is usually capable of detecting hCG by the first day of the missed menstrual period.

  • Simple-to-use - just one step with less than 30 seconds hands-on time
  • Easy-to-read results
  • Trust your results - no biotin interference, while detecting beta (β) subunit and other hCG variants
  • Get results quickly - results in as little as three minutes
  • Ready to use - includes transfer pipette for each test
  • QC Inside™ - two extra cassettes included at no additional charge to run external quality control testing for more confidence in your results

Recommended Storage

Store at room temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accuracy 100% (Urine), 99.5% (Serum)
Content And Storage Room Temperature
CPT Code 81025QW (Urine), 84703 (Serum)
Clia Complexity Waived (Urine), Non-waived (Serum)
Detectable Analytes hCG beta-core Fragments
Format Cassette
Sample Type Urine, Serum
Sensitivity 100%, 25 mIU/mL
Type Rapid Test
Includes 25 test devices, 25 disposable droppers, intructions for use (IFU). NOTE: two extra devices have been included in the kit for external QC testing.
Packaging Type Test Kit
Specificity 100% (Urine), 99% (Serum)
Test Time 3 to 5 min.
Analysis Time 3 to 5 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.